  • Search Results
  • Geniposide prevents rotenone-induced apoptosis in primary cultured neurons.
Geniposide prevents rotenone-induced apoptosis in primary cultured neurons.
Neural regeneration research (2015-12-23)
Lin Li, Juan Zhao, Ke Liu, Guang-Lai Li, Yan-Qing Han, Yue-Ze Liu
ABSTRACT
Geniposide, a monomer extracted from gardenia and widely used in Chinese medicine, is a novel agonist at the glucagon-like peptide-1 receptor. This receptor is involved in neuroprotection. In the present study, we sought to identify an anti-apoptotic mechanism for the treatment of neurodegenerative diseases. Primary cultured neurons were treated with different concentrations of rotenone for 48 hours. Morphological observation, cell counting kit-8 assay, lactate dehydrogenase detection and western blot assay demonstrated that 0.5 nM rotenone increased lactate dehydrogenase release, decreased the expression of procaspase-3 and Bcl-2, and increased cleaved caspase-3 expression in normal neurons. All these effects were prevented by geniposide. Our results indicate that geniposide diminished rotenone-induced injury in primary neurons by suppressing apoptosis. This may be one of the molecular mechanisms underlying the efficacy of geniposide in the treatment of neurodegenerative diseases.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%