  • Search Results
  • Targeting sphingosine kinase 2 (SphK2) by ABC294640 inhibits colorectal cancer cell growth in vitro and in vivo.
Targeting sphingosine kinase 2 (SphK2) by ABC294640 inhibits colorectal cancer cell growth in vitro and in vivo.
Journal of experimental & clinical cancer research : CR (2015-09-05)
Cai Xun, Min-Bin Chen, Li Qi, Zhang Tie-Ning, Xue Peng, Li Ning, Chen Zhi-Xiao, Wang Li-Wei
ABSTRACT
Colorectal cancer (CRC) is a major health problem in China and around the world. It is one of the leading causes of cancer-related deaths. Research groups are thus searching for novel and more efficient anti-CRC agents. Here we demonstrated that ABC294640, a novel SphK2 inhibitor, induced growth inhibition and apoptosis in transformed and primary CRC cells. The SphK activity was remarkably inhibited by ABC294640, accompanied by sphingosine-1-phosphate (S1P) depletion and ceramide incensement in CRC cells. Exogenously-added S1P inhibited ABC294640-induced HT-29 cell lethality. While C6 ceramide and SphK1 inhibitor SKI-II facilitated ABC294640-induced cytotoxicity against HT-29 cells. ABC294640 inhibited AKT-S6K1, but activated JNK signaling in transformed and primary CRC cells. JNK inhibitors (SP600125 and JNKi-II) alleviated ABC294640-induced CRC cell apoptosis. Moreover, a low concentration of ABC294640 sensitized the activity of 5-FU and cisplatin in vitro. In vivo, ABC294640 oral administration dramatically inhibited HT-29 xenografts growth in nude mice. Targeting of SphK2 by ABC294640 potently inhibits CRC cell growth both in vitro and in vivo, ABC294640 could be developed as a novel therapeutic for the treatment of CRC.
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
N-Hexanoyl-D-sphingosine, ≥98% (TLC), synthetic
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Liquified Phenol, ≥89.0%