  • Search Results
  • Anti-Human Immunodeficiency Virus Activity of Thiol-Ene Carbosilane Dendrimers and Their Potential Development as a Topical Microbicide.
Anti-Human Immunodeficiency Virus Activity of Thiol-Ene Carbosilane Dendrimers and Their Potential Development as a Topical Microbicide.
Journal of biomedical nanotechnology (2015-10-28)
Javier Sánchez-Rodríguez, Laura Díaz, Marta Galán, Marek Maly, Rafael Gómez, F Javier de la Mata, José L Jiménez, M Angeles Muñoz-Fernández
ABSTRACT
The concept of a "microbicide" was born out of the lack of a vaccine against HIV and the difficulty of women in ensuring the use of preventive prophylaxis by their partners, especially in developing countries. Approaches using polyanionic carbosilane dendrimers have shown promise in the development of new microbicides. We have developed and evaluated two anionic carbosilane dendrimers with sulfonate and carboxylate terminal groups, G2-STE16 and G2-CTE16. Both dendrimers showed high biosafety in human epithelial cell lines derived from the vagina and in primary blood human cells (PBMCs). The dendrimers not only have a greater capacity to block the entry of different X4- and R5-HIV-1 isolates into epithelial cells but also prevent the HIV-1 infection of activated PBMCs. The treatment of epithelial cells with different carbosilane dendrimers did not produce changes in the activation or proliferation of PBMCs or in the expression of CD4, CCR5 or CXCR4. Computational modeling showed significantly higher affinities for the complexes G2-STE16/gp120 and G2-CTE16/gp120. Moreover, no irritation or vaginal lesions were detected in female BALB/c mice after vaginal administration of the dendrimers. Summing up, G2-STE16 and G2-CTE16 are easy to synthesize and compatible with functional groups, and the purification steps are easy and short. Our results have clearly demonstrated that these dendrimers have high potency as a topical microbicide against HIV-1 infection.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Medroxyprogesterone
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)