MilliporeSigma
Combined Nurr1 and Foxa2 roles in the therapy of Parkinson's disease.
EMBO molecular medicine (2015-03-12)
Sang-Min Oh, Mi-Yoon Chang, Jae-Jin Song, Yong-Hee Rhee, Eun-Hye Joe, Hyun-Seob Lee, Sang-Hoon Yi, Sang-Hun Lee
ABSTRACT
Use of the physiological mechanisms promoting midbrain DA (mDA) neuron survival seems an appropriate option for developing treatments for Parkinson's disease (PD). mDA neurons are specifically marked by expression of the transcription factors Nurr1 and Foxa2. We show herein that Nurr1 and Foxa2 interact to protect mDA neurons against various toxic insults, but their expression is lost during aging and degenerative processes. In addition to their proposed cell-autonomous actions in mDA neurons, forced expression of these factors in neighboring glia synergistically protects degenerating mDA neurons in a paracrine mode. As a consequence of these bimodal actions, adeno-associated virus (AAV)-mediated gene delivery of Nurr1 and Foxa2 in a PD mouse model markedly protected mDA neurons and motor behaviors associated with nigrostriatal DA neurotransmission. The effects of the combined gene delivery were dramatic, highly reproducible, and sustained for at least 1 year, suggesting that expression of these factors is a promising approach in PD therapy.
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
L-Glutamine