  • Search Results
  • Mass spectrometry-based N-linked glycomic profiling as a means for tracking pancreatic cancer metastasis.
Mass spectrometry-based N-linked glycomic profiling as a means for tracking pancreatic cancer metastasis.
Carbohydrate research (2015-06-10)
Hae-Min Park, Mintai Peter Hwang, Yoon-Woo Kim, Kyoung-Jin Kim, Jang Mi Jin, Young Hwan Kim, Yung-Hun Yang, Kwan Hyi Lee, Yun-Gon Kim
ABSTRACT
The aberrant glycosylation profile on the surface of cancer cells has been recognized for its potential diagnostic value towards assessing tumor progression. In this study, we initially investigate N-glycan profiles on the surface of normal (HPDE) and cancerous (Capan-1, Panc-1, and MIA PaCa-2) pancreatic cell lines, which are from different sites of pancreatic tumor. The enzymatically deglycosylated total N-glycans are permethylated via a quantitative solid-phase method and then analyzed by using MALDI-TOF MS and MALDI-QIT-TOF MS. We demonstrate that the level of high-mannose type glycans is higher among Capan-1 cells-pancreatic cancer cells that have metastasized to the liver-than that observed among Panc-1 and MIA PaCa-2 cells-pancreatic cancer cells from the pancreas duct head and tail regions, respectively. Furthermore, the relative abundance of highly-branched sialyted N-glycans is significantly up-regulated on Panc-1 and MIA PaCa-2 pancreatic cancer cells compared to that of normal HPDE pancreas cells. Taken together, these results indicate that specific N-glycosylation profile changes in pancreatic cancer cells can be used to not only distinguish between normal and cancerous cells but also provide more information on their location and metastatic potential.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%