  • Search Results
  • Development of resistance to antiglioma agents in rat C6 cells caused collateral sensitivity to doxorubicin.
Development of resistance to antiglioma agents in rat C6 cells caused collateral sensitivity to doxorubicin.
Experimental cell research (2015-06-01)
Sonja Stojković, Ana Podolski-Renić, Jelena Dinić, Tijana Stanković, Jasna Banković, Stefan Hadžić, Verica Paunović, Aleksandra Isaković, Nikola Tanić, Milica Pešić
ABSTRACT
Chemoresistance is a severe limitation to glioblastoma (GBM) therapy and there is a strong need to understand the underlying mechanisms that determine its response to different chemotherapeutics. Therefore, we induced resistance in C6 rat glioma cell line, which considerably resembles the characteristics of human GBM. The resistant phenotype was developed by 3-bis (2-chloroethyl)-1-nitrosourea (BCNU), one of the most commonly used therapeutic drug in the course of GBM treatment. After confirmation of the cross-resistance to cisplatin (CPt) and temozolomide (TMZ) in newly established RC6 cell line, we examined cell death induction and DNA damage by these drugs. Resistance to apoptosis and deficiency in forming DNA double-strand breaks was followed by significant decrease in the mRNA expression of pro-apoptotic and anti-apoptotic genes. The development of drug resistance was associated with significant increase in reactive oxygen species (ROS) and decrease in oxidized to reduced gluthatione ratio in RC6 cell line indicating a reduced level of oxidative stress. The mRNA expression levels of manganese superoxid dismutase (MnSOD), inducible nitric oxide synthase (iNOS) and gluthatione peroxidase (GPx) were increased while hypoxia-inducible factor 1-α (HIF-1α) was decreased in RC6 compared to C6 cells. This was in line with obtained changes in ROS content and increased antioxidative capacity of RC6 cells. Importantly, RC6 cells demonstrated collateral sensitivity to doxorubicin (DOX). The analysis of this phenomenon revealed increased accumulation of DOX in RC6 cells due to their adaptation to high ROS content and acidification of cytoplasm. In conclusion, newly established RC6 rat glioma cell line could be used as a starting material for the development of allogenic animal model and preclinical evaluation of new antiglioma agents. Collateral sensitivity to DOX obtained after BCNU treatment may prompt new studies aimed to find efficient delivery of DOX to the glioma site in brain.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
trans-Platinum(II)diammine dichloride