  • Search Results
  • Dietary curcumin supplementation protects against heat-stress-impaired growth performance of broilers possibly through a mitochondrial pathway.
Dietary curcumin supplementation protects against heat-stress-impaired growth performance of broilers possibly through a mitochondrial pathway.
Journal of animal science (2015-05-29)
J F Zhang, Z P Hu, C H Lu, M X Yang, L L Zhang, T Wang
ABSTRACT
A total of 400 1-d-old Arbor Acres broiler chicks were raised at a recommended environmental temperature from d 1 to 20 (experimental day [ED] = ED1 to ED20). On ED21, the chicks were weighed and reallocated into 5 treatment groups, with 8 replicates of 10 birds each. The 5 treatment groups were as follows: the control group, in which chicks were housed at 22 ± 1°C and fed the basal diet, and the HS, HS-CUR50, HS-CUR100, and HS-CUR200 groups, in which chicks were housed at 34 ± 1°C for 8 h (0900-1700 h) and 22 ± 1°C for the rest time and fed the basal diet with 0, 50, 100, and 200 mg/kg curcumin, respectively. From ED21 to ED42, the heat treatment lasted for 20 consecutive days. The results showed that heat-stressed broilers had greater (P < 0.05) average head surface and rectal temperature on ED21 and ED42 than the non-heat-stressed broilers. Diets supplied with 50 and 100 mg/kg curcumin increased (P < 0.05) the G:F compared to the heat-stressed groups. Mitochondrial malondialdehyde levels, an index of lipid peroxidation, in the breast muscle were 15.15 and 9.09% higher (P < 0.05) in 50 and 100 mg/kg curcumin supplemented groups than that of the heat-stressed group, respectively. Curcumin supplementation (50, 100, and 200 mg/kg) increased (P < 0.05) mitochondrial glutathione content and glutathione peroxidase, glutathione S-transferase, and manganese superoxide dismutase activities compared to heat-stressed broilers. Curcumin supplementation (50, 100, and 200 mg/kg) resulted in a decrease (P < 0.05) of heat shock protein 70 mRNA levels in the breast muscle. The breast muscle mRNA expression of peroxisome proliferator-activated receptor γ coactivator 1α and nuclear respiratory factor 1 and 2 in heat-stressed groups was increased (P < 0.05) in response to dietary 100 mg/kg curcumin treatment. Additionally, when compared to the heat-stressed group, mitochondrial transcription factor A mRNA levels were increased (P < 0.05) by 17.64% in the 200 mg/kg curcumin supplemented group. In conclusion, dietary curcumin supplementation prevented heat-stress-impaired growth performance, possibly through improving the antioxidant defense system and enhancing the mitochondrial biogenesis.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free