  • Search Results
  • Quantitative determination of fucoidan using polyion-sensitive membrane electrodes.
Quantitative determination of fucoidan using polyion-sensitive membrane electrodes.
Analytica chimica acta (2015-05-24)
Ji Min Kim, Loc Nguyen, Mary Frances Barr, Michael Morabito, Damien Stringer, J Helen Fitton, Kelly A Mowery
ABSTRACT
The use of polyanion and polycation-sensitive membrane electrodes to detect five different preparations of fucoidan is described. Unlike linear polyanionic molecules previously measured with polymer membrane-based electrochemical sensors, fucoidans from marine brown algae are all highly branched, sulfated polysaccharides with varying charge densities and structures, depending on the species of seaweed, method of extraction used and extent of purification. When tridodecylmethylammonium (TDMA) was used as the ion-exchanger, a large, non-equilibrium EMF response was observed over a concentration range of 0.5-50 μg mL(-1) fucoidan. Fucoidan was also measured by titration with polycationic protamine, using a dinonylnaphthalene sulfonate (DNNS)-doped membrane electrode as the potentiometric endpoint detector. Potentiometric titration was used to determine the binding ratio between protamine and fucoidan at the neutralization endpoint for each fucoidan preparation. This binding ratio was then used to successfully determine the fucoidan content of commercially available nutritional supplements. Fucoidan was also measured in undiluted blood serum, demonstrating that this method may be applicable for measuring fucoidan for clinical applications.
Sigma-Aldrich
2-Nitrophenyl octyl ether, 99%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%