  • Search Results
  • Adjuvant Effect of an Alternative Plasticizer, Diisopropyl Adipate, on a Contact Hypersensitivity Mouse Model: Link with Sensory Ion Channel TRPA1 Activation.
Adjuvant Effect of an Alternative Plasticizer, Diisopropyl Adipate, on a Contact Hypersensitivity Mouse Model: Link with Sensory Ion Channel TRPA1 Activation.
Biological & pharmaceutical bulletin (2015-05-12)
Kohta Kurohane, Ayako Kimura, Rie Terasawa, Yurina Sahara, Kamiyu Kobayashi, Wakana Suzuki, Takeshi Matsuoka, Tatsuo Watanabe, Yasuyuki Imai
ABSTRACT
Due to health concerns about phthalate esters, the use of alternative plasticizers is being considered. Phthalate esters enhance skin sensitization to fluorescein isothiocyanate (FITC) in mouse models. We have demonstrated that phthalate esters stimulate transient receptor potential ankyrin 1 (TRPA1) cation channels expressed on sensory neurons. We also found a correlation between TRPA1 activation and the enhancing effect on FITC-induced contact hypersensitivity (CHS) when testing various types of phthalate esters. Here we investigated the effects of an alternative plasticizer, diisopropyl adipate (DIA). Activation of TRPA1 by DIA was demonstrated by calcium mobilization using Chinese hamster ovary cells expressing TRPA1 in vitro. The effect of DIA was inhibited by a TRPA1-specific antagonist, HC-030031. The presence of DIA or dibutyl phthalate (DBP; positive control) during skin sensitization of BALB/c mice to FITC augmented the CHS response, as revealed by the level of ear-swelling. The enhancing effect of DIA was inhibited by in vivo pretreatment with HC-030031. FITC-presenting CD11c(+) dendritic cell (DC)-trafficking to draining lymph nodes was facilitated both by DIA and by DBP. DBP and DIA were similarly active in the enhancement of interferon-γ production by draining lymph nodes, but the effect on interleukin-4 production was weaker with DIA. Overall, DIA activated TRPA1 and enhanced FITC-induced CHS, as DBP did. The adjuvant effects of adipate esters may need to be considered because they are used as ingredients in cosmetics and drug formulations topically applied to the skin.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HC-030031, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma