MilliporeSigma
TIPE2 protein prevents injury-induced restenosis in mice.
Biochimica et biophysica acta (2015-04-29)
Guizhong Zhang, Lianying Zhao, Yiyao Wang, Jie Shao, Jian Cui, Yunwei Lou, Minghong Geng, Na Zhang, Youhai H Chen, Suxia Liu
ABSTRACT
Proliferation of vascular smooth muscle cells (VSMCs) plays an important role in restenosis, a disease characterized by smooth muscle cell hyperplasia and neointimal formation. How proliferation signals are controlled to avoid restenosis is not fully understood. Here we report that TIPE2, the tumor necrosis factor (TNF) α-induced protein 8-like 2 (TNFAIP8L2), suppresses injury-induced restenosis by inhibiting VSMCs proliferation. TIPE2 was significantly upregulated in VSMCs in response to PDGF-BB stimuli and injury. Enforced TIPE2 expression significantly suppressed VSMCs proliferation and cell cycle progression, whereas TIPE2 deficiency in VSMCs promoted cell proliferation and upregulated the expression of Cyclins D1 and D3. TIPE2 likely regulated VSMC proliferation via Rac1-STAT3 and ERK1/2 signaling pathways. It blocked STAT3 activation and nuclear translocation in a Rac1-dependent manner. As a result, TIPE2-deficient VSMCs exhibited enhanced proliferation whereas TIPE2-deficient mice developed more severe restenosis in response to vascular injury. Conversely, adenovirus-mediated gene transfer of TIPE2 significantly reduced injury-induced restenosis in mice. These results indicate that TIPE2 plays a suppressive role in injury-induced restenosis and may serve as a new therapeutic target for treating the disease.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole, 98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)