  • Search Results
  • Construction of a novel phenol synthetic pathway in Escherichia coli through 4-hydroxybenzoate decarboxylation.
Construction of a novel phenol synthetic pathway in Escherichia coli through 4-hydroxybenzoate decarboxylation.
Applied microbiology and biotechnology (2015-03-12)
Liangtian Miao, Qingyan Li, Aipo Diao, Xueli Zhang, Yanhe Ma
ABSTRACT
Phenol is a bulk chemical with lots of applications in the chemical industry. Fermentative production of phenol had been realized in both Pseudomonas putida and Escherichia coli by recruiting tyrosine phenol-lyase (TPL). The TPL pathway needs tyrosine as the direct precursor for phenol production. In this work, a novel phenol synthetic pathway was created in E. coli by recruiting 4-hydroxybenzoate decarboxylase, which can convert 4-hydroxybenzoate to phenol and carbon dioxide. Activating 3-deoxy-D-arabino-heptulosonate-7-phosphate (DAHP) synthase and chorismate pyruvate lyase (UbiC) through plasmid overexpression led to 7- and 69-fold increase of phenol production, respectively, demonstrating that these two enzymes were the rate-limiting steps for phenol production. Genetically stable strains were then obtained by gene integration and gene modulation directly in chromosome. Phenol titer increased 147-fold (from 1.7 to 250 mg/L) after modulating the DAHP synthase, UbiC, and 4-hydroxybenzoate decarboxylase genes in chromosome. Five solvents were tested for two-phase extractive fermentation to eliminate phenol toxicity to E. coli cells. Tributyrin and dibutyl phthalate were the best two solvents for improving phenol production, leading to 23 and 30 % increase of total phenol production, respectively. Two-phase fed-batch fermentation of the best strain Phe009 was performed in a 7 L fermentor, which produced 9.51 g/L phenol with a yield of 0.061 g/g glucose.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology