  • Search Results
  • H9 Inhibits Tumor Growth and Induces Apoptosis via Intrinsic and Extrinsic Signaling Pathway in Human Non-Small Cell Lung Cancer Xenografts.
H9 Inhibits Tumor Growth and Induces Apoptosis via Intrinsic and Extrinsic Signaling Pathway in Human Non-Small Cell Lung Cancer Xenografts.
Journal of microbiology and biotechnology (2015-02-03)
Min-Je Kim, Sae-Bom Kwon, Seung Hoon Ham, Eui-Suk Jeong, Yang-Kyu Choi, Kang Duk Choi, Jin Tae Hong, Seung Hyun Jung, Do-Young Yoon
ABSTRACT
H9, a novel herbal extract, demonstrated cytotoxicity in A549 non-small cell lung cancer (NSCLC) cell lines. In this study, we investigated whether H9, and/or co-treatment with an anticancer drug, pemetrexed (PEM), inhibited tumor growth in BALB/c nude mice models bearing A549 NSCLC cells. The mice were separated into groups and administered H9 and PEM for 2 weeks. Protein and mRNA levels were detected using western blotting and reverse transcription polymerase chain reaction, respectively; immunohistochemistry (IHC) was also performed on the tumor tissues. H9 and co-treatment with PEM induced the cleavage of proapoptotic factors, such as caspase-3, caspase-8, caspase-9, and poly(ADP)-ribose polymerase (PARP). Expression levels of cell-death receptors involving Fas/FasL, TNF-related apoptosisinducing ligands (TRAIL), and TRAIL receptors were increased by H9 and co-treatment with PEM. Furthermore, analysis of levels of cell-cycle modulating proteins indicated that tumor cells were arrested in the G1/S phase. In addition, the phosphatidylinositol-4,5-bisphosphate 3-kinase (PI3K)/Akt survival signaling pathways were inhibited by H9 and co-treatment with PEM. In conclusion, H9 and co-treatment with PEM inhibited tumor growth in BALB/c nude mice models bearing A549 NSCLC cells. These results indicate that H9 and co-treatment with PEM can be used as an anticancer therapy in NSCLC.
Product Description
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Aluminum sulfate, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium iodate, ≥99%
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri