  • Search Results
  • Xylan-degrading enzymes from Aspergillus terreus: Physicochemical features and functional studies on hydrolysis of cellulose pulp.
Xylan-degrading enzymes from Aspergillus terreus: Physicochemical features and functional studies on hydrolysis of cellulose pulp.
Carbohydrate polymers (2015-10-03)
Leonora Rios de Souza Moreira, Alice da Cunha Morales Álvares, Francides Gomes da Silva, Sonia Maria de Freitas, Edivaldo Ximenes Ferreira Filho
ABSTRACT
Two endo-β-1,4-xylanases named XylT1 and XylT2, previously purified from Aspergillus terreus, were structurally investigated by fluorescence quenching and characterized with respect to their binding properties with phenolic compounds. Neutral and charged quenchers had access to both enzymes in neutral and alkaline pHs. The greatest access was noted for the negative quencher, possibly due to positive amino acid residues in the vicinity of tryptophan. These tryptophan environments may partially explain the conformational differences and lower binding constants of phenolic compounds for XylT2 than XylT1Phenolic compounds had lower binding constants for XylT2 than XylT1. These results show that xylanases present structural and functional differences, despite belonging to similar families. XylT1 and XylT2 were also evaluated for their ability to hydrolyze cellulose pulp in different stages of bleaching. Both enzymes promoted hydrolysis of cellulose pulps, which was confirmed by the release of total reducing sugars, pentoses and chromophoric material. Analysis of released xylooligosaccharides demonstrated a preferential release of xylobiose. None of xylanases released glucose, showing that they do not hydrolyze the cellulose present in the pulp, making both enzymes excellent choices for bio-bleaching applications.
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)