  • Search Results
  • Shape and surface chemistry effects on the cytotoxicity and cellular uptake of metallic nanorods and nanospheres.
Shape and surface chemistry effects on the cytotoxicity and cellular uptake of metallic nanorods and nanospheres.
Journal of biomedical materials research. Part A (2015-06-09)
Pelagie Marlene Favi, Mariana Morales Valencia, Paul Robert Elliott, Alejandro Restrepo, Ming Gao, Hanchen Huang, Juan Jose Pavon, Thomas Jay Webster
ABSTRACT
Metallic nanoparticles (such as gold and silver) have been intensely studied for wound healing applications due to their ability to be easily functionalized, possess antibacterial properties, and their strong potential for targeted drug release. In this study, rod-shaped silver nanorods (AgNRs) and gold nanorods (AuNRs) were fabricated by electron beam physical vapor deposition (EBPVD), and their cytotoxicity toward human skin fibroblasts were assessed and compared to sphere-shaped silver nanospheres (AgNSs) and gold nanospheres (AuNSs). Results showed that the 39.94 nm AgNSs showed the greatest toxicity with fibroblast cells followed by the 61.06 nm AuNSs, ∼556 nm × 47 nm (11.8:1 aspect ratio) AgNRs, and the ∼534 nm × 65 nm (8.2:1 aspect ratio) AuNRs demonstrated the least amount of toxicity. The calculated IC50 (50% inhibitory concentration) value for the AgNRs exposed to fibroblasts was greater after 4 days of exposure (387.3 μg mL(-1)) compared to the AgNSs and AuNSs (4.3 and 23.4 μg mL(-1), respectively), indicating that these spherical metallic nanoparticles displayed a greater toxicity to fibroblast cells. The IC50 value could not be measured for the AuNRs due to an incomplete dose response curve. The reduced cell toxicity with the presently developed rod-shaped nanoparticles suggests that they may be promising materials for use in numerous biomedical applications.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)