  • Search Results
  • Effects of dietary supplementation with fermented ginkgo leaves on antioxidant capacity, intestinal morphology and microbial ecology in broiler chicks.
Effects of dietary supplementation with fermented ginkgo leaves on antioxidant capacity, intestinal morphology and microbial ecology in broiler chicks.
British poultry science (2015-04-15)
X H Zhang, Z Y Sun, F L Cao, H Ahmad, X H Yang, L G Zhao, T Wang
ABSTRACT
1. The purpose of this study was to evaluate the effects of supplementing diets with three types of fermented Ginkgo-leaves (FGL) on growth, antioxidant capacity, intestinal morphology and microbial ecology in broiler chicks. 2. A total of 300 d-old broilers were randomly allocated to 4 dietary treatments with 6 replications of 10 birds each. Birds were fed on basal diets (Control) or basal diets supplemented with 0.5% FGL with Candida utilis (CF group), Aspergillus niger (AF group) or their combined fermentation (CAF group), respectively, for a 42 d feeding trial. 3. AF and CAF supplementation improved body weight gain (BWG) (22-42 d) and feed conversion ratio (22-42 d and 1-42 d). Concentrations of serum α-tocopherol in CAF group, as well as hepatic α-tocopherol in the three FGL groups were increased, while hepatic reactive oxygen species (ROS) levels were greatly decreased in group AF and CAF. Chickens in AF and CAF groups had decreased hepatic protein carbonyls and malondialdehyde (MDA), as well as jejunal and ileal protein carbonyls. The total superoxide dismutase (T-SOD) activities and glutathione (GSH) of both jejunum and ileum of the CAF group were higher than the other groups. 4. Duodenal and jejunal villous height of birds fed on the AF and CAF diets were increased, while jejunal crypt depth (CD) was decreased. Furthermore, birds fed on AF and CAF supplemented diets had increased ileal lactobacilli populations. Decreased ileal and caecal Escherichia coli and Salmonellas populations was found for the birds fed on CAF supplemented diets. 5. The present study may indicate that the improved feed efficiency and intestinal functions in the group supplemented with AF and CAF are directly connected with the improved antioxidant capacity and intestinal microbial ecology.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous