  • Search Results
  • Proteomic analyses of membrane enriched proteins of Leishmania donovani Indian clinical isolate by mass spectrometry.
Proteomic analyses of membrane enriched proteins of Leishmania donovani Indian clinical isolate by mass spectrometry.
Parasitology international (2015-01-20)
Awanish Kumar, Pragya Misra, Brijesh Sisodia, Ajit Kumar Shasany, Shyam Sundar, Anuradha Dube
ABSTRACT
Visceral leishmaniasis (VL) is a major fatal disease prevalent in North-East India, caused by a protozoan parasite Leishmania donovani. The parasite multiplies and thrives inside mammalian macrophages and is transmitted by the bite of the sandfly. Due to the unsatisfactory treatment measures, increasing drug resistance and the advent of HIV-Leishmania co-infection there has been an urgent need to develop novel drug/vaccine targets against VL. Target identification is the key step in drug discovery and proteomics seems to be a suitable strategy for it due to the availability of Leishmania major, Leishmania infantum, Leishmania braziliensis, Leishmania donovani, Leishmania mexicana and Leishmania tarentolae genome sequence. Since, majority of proteome analyses of Leishmania have, so far, been performed on whole-cell extracts; this study is dealing with the sub-proteome analysis of the membrane-enriched protein (MEP) fractions of L. donovani. The analysis of 95 protein spots of the MEPs from two dimensional (2-D) gel image through matrix asserted laser desorption ionization-time of flight/mass spectrometry (MALDI-TOF/MS) endorsed the identification of various relevant functional proteins. Out of 95 the MEP spots 72 have been identified and were classified on the basis of their biological function. Several proteins of unknown function that belong to different classes like cell signaling, transmembrane receptors, and transporters have been identified which could be the new potential therapeutic targets against VL in future. The proteome array of the MEPs contributes to further elucidation of the biological system of L. donovani as well as host-parasite relationships which may be further investigated for their crucial biological role in L. donovani for disease management.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%