  • Search Results
  • One-pot synthesis of multifunctional Au@graphene oxide nanocolloid core@shell nanoparticles for Raman bioimaging, photothermal, and photodynamic therapy.
One-pot synthesis of multifunctional Au@graphene oxide nanocolloid core@shell nanoparticles for Raman bioimaging, photothermal, and photodynamic therapy.
Small (Weinheim an der Bergstrasse, Germany) (2015-01-30)
Young-Kwan Kim, Hee-Kyung Na, Seongchan Kim, Hongje Jang, Sung-Jin Chang, Dal-Hee Min
ABSTRACT
The paper reports a facile one-pot synthesis of core@shell nanoparticles (NPs) composed of Au core and graphene oxide nanocolloid (GON) shell. Unique properties of Au NPs and GON can be incorporated into a single nanohybrid structure to provide desirable functions for theranosis such as localized surface plasmon resonance, Raman scattering, amphiphilic surface, and photothermal conversion. Synthesis of Au@GON NPs is achieved by simple one-pot reaction in aqueous phase utilizing GON as a reducing and stabilizing agent without any additional reducing agent. The zinc phthalocyanine, a photosensitizer, loaded Au@GON NPs show excellent multifunctional properties for combinational treatment of photothermal and photodynamic therapy in addition to Raman bioimaging with low cytotoxicity.
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%