  • Search Results
  • Identification of a sustained neurogenic zone at the dorsal surface of the adult mouse hippocampus and its regulation by the chemokine SDF-1.
Identification of a sustained neurogenic zone at the dorsal surface of the adult mouse hippocampus and its regulation by the chemokine SDF-1.
Hippocampus (2015-02-07)
Abdelhak Belmadani, Dongjun Ren, Bula J Bhattacharyya, Katharina B Rothwangl, Thomas J Hope, Harris Perlman, Richard J Miller
ABSTRACT
We identified a previously unknown neurogenic region at the dorsal surface of the hippocampus; (the "subhippocampal zone," SHZ) in the adult brain. Using a reporter mouse in which SHZ cells and their progeny could be traced through the expression of EGFP under the control of the CXCR4 chemokine receptor promoter we observed the presence of a pool of EGFP expressing cells migrating in direction of the dentate gyrus (DG), which is maintained throughout adulthood. This population appeared to originate from the SHZ where cells entered a caudal migratory stream (aCMS) that included the fimbria, the meninges and the DG. Deletion of CXCR4 from neural stem cells (NSCs) or neuroinflammation resulted in the appearance of neurons in the DG, which were the result of migration of NSCs from the SHZ. Some of these neurons were ectopically placed. Our observations indicate that the SHZ is a neurogenic zone in the adult brain through migration of NSCs in the aCMS. Regulation of CXCR4 signaling in these cells may be involved in repair of the DG and may also give rise to ectopic granule cells in the DG in the context of neuropathology.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)