  • Search Results
  • Loop-mediated isothermal amplification as alternative to PCR for the diagnosis of extra-pulmonary tuberculosis.
Loop-mediated isothermal amplification as alternative to PCR for the diagnosis of extra-pulmonary tuberculosis.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease (2015-07-15)
D Joon, M Nimesh, D Saluja
ABSTRACT
The main challenge in combatting extra-pulmonary tuberculosis (EPTB) is the lack of a rapid, reliable and inexpensive diagnostic test for the detection of Mycobacterium tuberculosis. To evaluate the diagnostic potential of an L-serine dehydratase gene (sdaA) loop-mediated isothermal amplification (LAMP) assay for the detection of M. tuberculosis in clinical specimens from presumptive EPTB patients. An in-house sdaA LAMP assay was used to analyse clinical specimens (n = 315) for the diagnosis of EPTB compared with culture and the composite reference standard (CRS) comprising culture and polymerase chain reaction (PCR) using insertion sequence (IS) 6110 and mpb64 as target genes. The sdaA LAMP assay showed the highest sensitivity (93.3%) in comparison to culture; the sensitivity of IS6110 PCR, mpb64 and sdaA PCR assay was respectively 80%, 86.7% and 90%. In comparison to CRS, the LAMP assay had a sensitivity of 92.5% and a specificity of 99.2%, with a high positive (121.11) and a low negative likelihood ratio (0.08). Due to its speed, simplicity, sensitivity and specificity, the sdaA LAMP assay is a potential diagnostic test for the diagnosis of EPTB, particularly in resource-limited settings.
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, meets EP, USP testing specifications
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)