  • Search Results
  • Network nanostructured polypyrrole hydrogel/Au composites as enhanced electrochemical biosensing platform.
Network nanostructured polypyrrole hydrogel/Au composites as enhanced electrochemical biosensing platform.
Scientific reports (2015-06-16)
Qinfeng Rong, Hongliang Han, Feng Feng, Zhanfang Ma
ABSTRACT
In this work, a new network nanocomposite composed of polypyrrole hydrogel (PPy hydrogel) loaded gold nanoparticles (AuNPs) was prepared. The PPy hydrogel was directly synthesized by mixing the pyrrole monomer and phytic acid, and the mixed solution can be gelated to form hydrogel at once. The three-dimensional network nanostructured PPy hydrogel not only provided a greater effective surface area for increasing the quantity of immobilized biomolecules and facilitated the transport of electrons and ions, but also exhibited an improved conductivity. Meanwhile, the electrodeposited AuNPs on the PPy hydrogel can further increase the specific surface area to capture a large amount of antibodies as well as improve the capability of electron transfer. The network PPy hydrogel/Au nanocomposites were successfully employed for the fabrication of a sensitive label-free amperometric immunosensor. Carcinoembryonic antigen (CEA) was used as a model protein. The proposed immunosensor exhibited a wide linear detection range from 1 fg mL(-1) to 200 ng mL(-1), and an ultralow limit of detection of 0.16 g mL(-1) (S/N = 3), and it also possessed good selectivity. Moreover, the detection of CEA in ten human serums showed satisfactory accuracy compared with the data determined by ELISA, indicating that the immunosensor provided potential application for clinical diagnosis.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)