  • Search Results
  • Assay of ceftazidime and cefepime based on fluorescence quenching of carbon quantum dots.
Assay of ceftazidime and cefepime based on fluorescence quenching of carbon quantum dots.
Luminescence : the journal of biological and chemical luminescence (2015-02-27)
Yu Huang, Ying Zhang, Zhengyu Yan, Shenghua Liao
ABSTRACT
A novel and sensitive method for the determination of ceftazidime and cefepime in an active pharmaceutical ingredient (API) has been developed based on the fluorescence quenching of poly(ethylene glycol) (PEG)2000-capped carbon quantum dots (CQDs) prepared using a chemical oxidation method. The quenching of fluorescence intensity is proportional to the concentration of ceftazidime and cefepime over the range of 0.33-3.30 and 0.24-2.40 µg/mL, respectively. The mode of interaction between PEG2000-capped CQDs and ceftazidime/cefepime in aqueous solutions was investigated using a fluorescence, UV/Vis and Fourier transform infrared spectrometry (FTIR) at physiological pH. UV/Vis and FTIR spectra demonstrated that ground state compounds were formed through hydrophobic interaction the fluorescence quenching of CQDs caused by ceftazidime and cefepime. The quenching constants decreased with increases in temperature, which was consistent with static quenching.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%