  • Search Results
  • Factors other than metalloprotease are required for full virulence of French Vibrio tubiashii isolates in oyster larvae.
Factors other than metalloprotease are required for full virulence of French Vibrio tubiashii isolates in oyster larvae.
Microbiology (Reading, England) (2015-02-24)
Rachida Mersni-Achour, Yosra Ben Cheikh, Vianney Pichereau, Ibtissem Doghri, Cédric Etien, Lionel Dégremont, Denis Saulnier, Ingrid Fruitier-Arnaudin, Marie-Agnès Travers
ABSTRACT
Vibrio tubiashii is a marine pathogen isolated from larval and juvenile bivalve molluscs that causes bacillary necrosis. Recent studies demonstrated the isolation of this species in a French experimental hatchery/nursery affecting Crassostrea gigas spat in 2007. Here, using larvae of C. gigas as an interaction model, we showed that the French V. tubiashii is virulent to larvae and can cause bacillary necrosis symptoms with an LD50 of about 2.3 × 10(3) c.f.u. ml(-1) after 24 h. Moreover, complete or gel permeation HPLC fractionated extracellular products (ECPs) of this strain appeared toxic to larvae. MS-MS analysis of the different ECP fractions revealed the existence of an extracellular metalloprotease and other suspected virulence factors. This observation is also supported by the expression level of some potential virulence factors. The overall results suggest that the pathology caused by the French V. tubiashii in C. gigas oysters is caused by a group of toxic factors and not only the metalloprotease.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)