  • Search Results
  • Perinatal exposure to low-dose of bisphenol A causes anxiety-like alteration in adrenal axis regulation and behaviors of rat offspring: a potential role for metabotropic glutamate 2/3 receptors.
Perinatal exposure to low-dose of bisphenol A causes anxiety-like alteration in adrenal axis regulation and behaviors of rat offspring: a potential role for metabotropic glutamate 2/3 receptors.
Journal of psychiatric research (2015-03-31)
Rong Zhou, Fang Chen, Xuejiao Feng, Libin Zhou, Yingchun Li, Ling Chen
ABSTRACT
The present study focuses on detecting anxiety-like behavior and associated neurochemical alterations in adolescent rats exposed perinatally to bisphenol A (BPA), an estrogen-mimicking endocrine disrupter and investigating the possible involvement of metabotropic glutamate 2/3 receptors (mGlu2/3 receptors) in BPA-induced anxiogenic effects. When female breeders were administered orally with BPA (40 μg/kg/d) during pregnancy and lactation, their pups (here named 'BPA-exposed offspring') developed an anxiety-like phenotype, characterized by the hyperactivity of the hypothalamic-pituitary-adrenal (HPA) axis, impaired glucocorticoid receptor (GR)-mediated negative feedback regulation of the HPA axis, altered hippocampal synaptic plasticity and increased anxiety-like behaviors. BPA-exposed offspring also showed a reduced expression of mGlu2/3 receptors in the hippocampus. BPA-exposed offspring further subjected to systemic administration of mGlu2/3 receptor agonist (LY379268, 0.5 mg/kg, i.p.) or antagonist (LY341495, 1.5 mg/kg, i.p.) twice per day for 6 days. The results indicated that chronic LY379268 treatment corrected the anxiety-like behaviors and associated neurochemical and endocrinological alterations in BPA-exposed offspring. Our data demonstrate for the first time that the perinatal BPA exposure induces an anxiety-like phenotype in behaviors and -related neuroendocrinology, and suggest that the changes in mGlu2/3 receptor might lie at the core of the pathological reprogramming triggered by early-life adversity. mGlu2/3 receptor may serve as a novel biomarker and potential therapeutic target for anxiety disorders associated with adverse early-life agents including perinatal BPA exposure.
Sigma-Aldrich
Dexamethasone 21-phosphate disodium salt, ≥98%
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%