  • Search Results
  • The role of nucleotide-binding oligomerization domain 1 during cytokine production by macrophages in response to Mycobacterium tuberculosis infection.
The role of nucleotide-binding oligomerization domain 1 during cytokine production by macrophages in response to Mycobacterium tuberculosis infection.
Immunobiology (2015-08-10)
Jun-Young Lee, Eun-Ha Hwang, Dong-Jae Kim, Sang-Muk Oh, Kyung-Bok Lee, Sung Jae Shin, Jong-Hwan Park
ABSTRACT
Tuberculosis due to Mycobacterium tuberculosis infection is a leading cause of death worldwide. Recognition of this pathogen is crucial for the activation of innate and adaptive immune responses. Nucleotide-binding oligomerization domain (Nod)1 and Nod2 are cytoplasmic receptors that can detect unique muropeptides of bacterial peptidoglycan. Nod2 is critical for the initiation of the host immune response against M. tuberculosis infection, however the role of Nod1 remains largely unknown. We investigated the role of Nod1 with respect to cytokine production by bone marrow-derived macrophages (BMDMs) in response to M. tuberculosis infection. Production of proinflammatory cytokines, such as IL-6, TNF-α, and IL-1β were induced in BMDMs; cytokine levels were not affected by a deficiency in Nod1. Activation of NF-κB and MAPKs was also comparable between wild-type and Nod1-deficient BMDMs. Levels of IL-6 and IL-1β were reduced in Nod1/Nod2 double-deficient BMDMs to a greater extent than in Nod2-deficient cells. Furthermore, when signaling of Toll-like receptors (TLRs) was inhibited by lipopolysaccharide pre-treatment, cytokine production was diminished in Nod1-deficient BMDMs. Our results indicate that Nod1 cooperates with Nod2 or TLRs to produce cytokines in macrophages in response to M. tuberculosis infection.
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)