  • Search Results
  • Effects of scleral cross-linking using genipin on the process of form-deprivation myopia in the guinea pig: a randomized controlled experimental study.
Effects of scleral cross-linking using genipin on the process of form-deprivation myopia in the guinea pig: a randomized controlled experimental study.
BMC ophthalmology (2015-07-30)
Mengmeng Wang, Christine Carole C Corpuz
ABSTRACT
Scleral cross-linking (CXL) is a novel attempt to slow down the axial elongation process in animal eyes. As a natural CXL reagent, genipin would be also effective for the prevention of myopia process. Thus, the present study was designed to evaluate the effects of scleral cross-linking using genipin on the form-deprivation (FD) myopia process of guinea pigs. Twenty-seven 3-week-old pigmented guinea pigs were randomly divided into three groups. Group A (n = 8) is the untreated control group. Group B (n = 8) is the FD control group, where all eyes were induced with monocular FD for 21 days. In Group C (n = 11), a sub-Tenon injection of 0.10 mL 0.50 % genipin was performed on FD eyes at day 0, 7 and 14 during the 21-day monocular FD. The ocular refraction, axial length, biomechanical test and light and electron microscopy were measured on all eyes to check the efficacy and safety of this scleral CXL technique. Compared with Group A, significant increases in myopic refractive errors, axial elongation and reductions of scleral fibril diameter and density were observed in the 21-day FD eyes of Group B (P < 0.05). In Group C, the scleral CXL resulted in less myopia and axial elongation as compared with Group B (P < 0.05); a significant thickening of scleral fibrils was found after sub-Tenon injections of genipin; no histological damage on the retina or choroid was observed in Group C at the end of this study. The FD myopia in guinea pig eyes was effectively blocked by the scleral CXL using sub-Tenon injections of genipin. No histological damage was found on the retina or choroid of these treated eyes. Further studies are needed to examine the long-term efficacy and safety of this CXL technique.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC