  • Search Results
  • Nanoshell-mediated targeted photothermal therapy of HER2 human breast cancer cells using pulsed and continuous wave lasers: an in vitro study.
Nanoshell-mediated targeted photothermal therapy of HER2 human breast cancer cells using pulsed and continuous wave lasers: an in vitro study.
Lasers in medical science (2015-07-04)
Mohammad E Khosroshahi, Zahra Hassannejad, Masoumeh Firouzi, Ahmad R Arshi
ABSTRACT
In this study, we report the apoptosis induction in HER2 overexpressed breast cancer cells using pulsed, continuous wave lasers and polyvinylpyrrolidone (PVP)-stabilized magneto-plasmonic nanoshells (PVP-MPNS) delivered by immunoliposomes. The immunoliposomes containing PVP-MPNS were fabricated and characterized. Heating efficiency of the synthesized nanostructures was calculated. The effect of functionalization on cellular uptake of nanoparticles was assessed using two cell lines of BT-474 and Calu-6. The best uptake result was achieved by functionalized liposome (MPNS-LAb) and BT-474. Also, the interaction of 514 nm argon (Ar) and Nd/YAG second harmonic 532-nm lasers with nanoparticles was investigated based on the temperature rise of the nanoshell suspension and the release value of 5(6)-carboxyfluorescein (CF) from CF/MPNS-loaded liposomes. The temperature increase of the suspensions after ten consecutive pulses of 532 nm and 5 min of irradiation by Ar laser were measured approximately 2 and 12 °C, respectively. The irradiation of CF/MPNS-loaded liposomes by Ar laser for 3 min resulted in 24.3 % release of CF, and in the case of 532 nm laser, the release was laser energy dependent. Furthermore, the comparison of CF release showed a higher efficiency for the Ar laser than by direct heating of nanoshell suspension using circulating water. The percentage of cell apoptosis after irradiation by Ar and 532 nm lasers were 44.6 and 42.6 %, respectively. The obtained results suggest that controlling the NP-laser interaction using optical properties of nanoshells and the laser parameters can be used to develop a new cancer therapy modality via targeted nanoshell and drug delivery.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, Plant-Derived
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
5(6)-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis