  • Search Results
  • N-trimethyl chitosan chloride-coated PLGA nanoparticles overcoming multiple barriers to oral insulin absorption.
N-trimethyl chitosan chloride-coated PLGA nanoparticles overcoming multiple barriers to oral insulin absorption.
ACS applied materials & interfaces (2015-06-26)
Jianyong Sheng, Limei Han, Jing Qin, Ge Ru, Ruixiang Li, Lihong Wu, Dongqi Cui, Pei Yang, Yuwei He, Jianxin Wang
ABSTRACT
Although several strategies have been applied for oral insulin delivery to improve insulin bioavailability, little success has been achieved. To overcome multiple barriers to oral insulin absorption simultaneously, insulin-loaded N-trimethyl chitosan chloride (TMC)-coated polylactide-co-glycoside (PLGA) nanoparticles (Ins TMC-PLGA NPs) were formulated in our study. The Ins TMC-PLGA NPs were prepared using the double-emulsion solvent evaporation method and were characterized to determine their size (247.6 ± 7.2 nm), ζ-potential (45.2 ± 4.6 mV), insulin-loading capacity (7.8 ± 0.5%) and encapsulation efficiency (47.0 ± 2.9%). The stability and insulin release of the nanoparticles in enzyme-containing simulated gastrointestinal fluids suggested that the TMC-PLGA NPs could partially protect insulin from enzymatic degradation. Compared with unmodified PLGA NPs, the positively charged TMC-PLGA NPs could improve the mucus penetration of insulin in mucus-secreting HT29-MTX cells, the cellular uptake of insulin via clathrin- or adsorption-mediated endocytosis in Caco-2 cells and the permeation of insulin across a Caco-2 cell monolayer through tight junction opening. After oral administration in mice, the TMC-PLGA NPs moved more slowly through the gastrointestinal tract compared with unmodified PLGA NPs, indicating the mucoadhesive property of the nanoparticles after TMC coating. Additionally, in pharmacological studies in diabetic rats, orally administered Ins TMC-PLGA NPs produced a stronger hypoglycemic effect, with 2-fold higher relative pharmacological availability compared with unmodified NPs. In conclusion, oral insulin absorption is improved by TMC-PLGA NPs with the multiple absorption barriers overcome simultaneously. TMC-PLGA NPs may be a promising drug delivery system for oral administration of macromolecular therapeutics.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems