  • Search Results
  • Chemical conversion of self-assembled hexadecyl monolayers with active oxygen species generated by vacuum ultraviolet irradiation in an atmospheric environment.
Chemical conversion of self-assembled hexadecyl monolayers with active oxygen species generated by vacuum ultraviolet irradiation in an atmospheric environment.
Soft matter (2015-06-18)
Ahmed I A Soliman, Takashi Ichii, Toru Utsunomiya, Hiroyuki Sugimura
ABSTRACT
Vacuum ultraviolet (VUV, λ = 172 nm) irradiation of alkyl self-assembled monolayers (SAMs) in the presence of dry air alters their surface properties. In this work, UV photochemically prepared hexadecyl (HD)-SAMs on hydrogen-terminated silicon substrates were irradiated by VUV light in dry air, which generated active oxygen species upon excitation of the atmospheric oxygen molecules. These active oxygen species converted the terminal methyl groups of the SAMs to polar functional groups, which were examined quantitatively by X-ray photoelectron spectroscopy (XPS) and chemical labeling. At the first stage of VUV irradiation, the surface of SAMs was functionalized, and the ratios of the generated polar functional groups markedly increased. With the elongation of the irradiation period, the SAMs gradually degraded, and the total polar group percentages gradually decreased. The difference between the oxygenated carbon components derived by the deconvolution of the XPS carbon (C1s) spectrum and the chemical labeling of polar groups revealed enormous quantities of ethereal and ester groups that cannot react with the labeling reagents but are included in the C1s spectral envelope. These modifications were reflected on morphological structures of SAMs, which were gradually distorted until a complete amorphous structure was obtained after the complete elimination of HD-SAMs.
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
1-Hexadecene, ≥98.5%
Sigma-Aldrich
4-Methylumbelliferyl β-D-glucopyranoside, β-glucosidase substrate
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%