  • Search Results
  • Growth inhibitory and chemo-sensitization effects of naringenin, a natural flavanone purified from Thymus vulgaris, on human breast and colorectal cancer.
Growth inhibitory and chemo-sensitization effects of naringenin, a natural flavanone purified from Thymus vulgaris, on human breast and colorectal cancer.
Cancer cell international (2015-06-16)
Mohamed Salah I Abaza, Khaled Y Orabi, Ebtehal Al-Quattan, Raja'a J Al-Attiyah
ABSTRACT
Natural products with diverse bioactivities are becoming an important source of novel agents with medicinal potential. Cancer is a devastating disease that causes the death of millions of people each year. Thus, intense research has been conducted on several natural products to develop novel anticancer drugs. Chromatographic and spectral techniques were used for the isolation and identification of naringenin (Nar). MTT, flow cytometry, western blotting, Real Time PCR were used to test anticancer and chemosensitizing effects of Nar, cell cycle, apoptosis, and expression of cell cycle, apoptosis, pro-survival and anti-survival-related genes. In the present study, Thymus vulgaris ethanol extract was purified repeatedly to produce several compounds including the known flavanone, Nar which was identified using different spectral techniques. Nar was shown to inhibit both human colorectal and breast cancer cell growth in a dose- and time-dependent manner through cell cycle arrest at S- and G2/M-phases accompanied by an increase in apoptotic cell death. Additionally, Nar altered the expression of apoptosis and cell-cycle regulatory genes by down-regulating Cdk4, Cdk6, Cdk7, Bcl2, x-IAP and c-IAP-2 and up-regulating p18, p19, p21, caspases 3, 7, 8 and 9, Bak, AIF and Bax in both colorectal and breast cancer cells. Conversely, it diminished the expression levels of the cell survival factors PI3K, pAkt, pIκBα and NFκBp65. Moreover, Nar enhanced the sensitivity of colorectal and breast cancer cells to DNA-acting drugs. These findings provide evidence that Nar's pro-apoptotic and chemo-sensitizing effects are mediated by perturbation of cell cycle, upregulation of pro-apoptotic genes and down-regulation of anti-apoptotic genes and inhibition of pro-survival signaling pathways. In conclusion, Nar might be a promising candidate for chemoprevention and/or chemotherapy of human cancers. However, further studies exploring this therapeutic strategy are necessary.
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
HEPES
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder