  • Search Results
  • A novel model of megavoltage radiation-induced oral mucositis in hamsters: Role of inflammatory cytokines and nitric oxide.
A novel model of megavoltage radiation-induced oral mucositis in hamsters: Role of inflammatory cytokines and nitric oxide.
International journal of radiation biology (2015-03-12)
José Fernando Bastos Moura, José Maurício Segundo Correia Mota, Caio Abner Vitorino Leite, Deysi Viviana T Wong, Nilfácio Prado Bezerra, Gerly Anne De Castro Brito, Vilma Lima, Fernando Queiroz Cunha, Ronaldo Albuquerque Ribeiro
ABSTRACT
To design a novel model to study Cobalt-60 (Co-60)-induced radiation mucositis and to describe the pathways involved in its development. Hamsters' cheeks were treated with Co-60 radiation (10, 20, 30 or 35 Gy). Three days later, oral mucosa scarification was performed with a needle. The animals were euthanized at day 13 (D + 13) after irradiation. Gross and microscopic alterations were evaluated by a new score system that we developed. Also, neutrophil infiltration, tumor necrosis factor-α (TNF-α), interleukin (IL)-1β, IL-10, inducible nitric oxide synthase (iNOS), nitric oxide (NO) and nitrite were assessed in oral mucosa. We also tried to establish the roles of TNF-α and IL-1β and iNOS in our model using pharmacological approaches with pentoxiphylline (PTX) and aminoguanidine (AMG), respectively. We found that a single administration of 35 Gy of Co-60, followed by mechanical scratches 3 days later, induced oral mucositis in hamsters. Animals with mucositis lost weight and had a survival median of 13 days, the time at which peak inflammation occurs. We noticed increased levels of NO, iNOS, TNF-α and IL-1β and a reduced concentration of IL-10. PTX partially prevented the mucositis phenotype by reducing the levels of inflammatory mediators and iNOS expression. Additionally, AMG, a selective inhibitor of iNOS, reduced Co-60-induced oral mucositis through reducing NO production. We described a novel model of megavoltage radiation-induced oral mucositis in hamsters. TNF-α, IL-1β and NO seem to play a role in the pathophysiology of this model.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Sigma-Aldrich
Chloral hydrate, meets analytical specification of Ph. Eur., BP, USP, 99.5-101%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
Chloral hydrate
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
SAFC
Hexadecyltrimethylammonium bromide