  • Search Results
  • Renal developmental defects resulting from in utero hypoxia are associated with suppression of ureteric β-catenin signaling.
Renal developmental defects resulting from in utero hypoxia are associated with suppression of ureteric β-catenin signaling.
Kidney international (2015-01-15)
Lorine J Wilkinson, Cailda S Neal, Reetu R Singh, Duncan B Sparrow, Nyoman D Kurniawan, Adler Ju, Stuart M Grieve, Sally L Dunwoodie, Karen M Moritz, Melissa H Little
ABSTRACT
Gestational stressors, including glucocorticoids and protein restriction, can affect kidney development and hence final nephron number. Since hypoxia is a common insult during pregnancy, we studied the influence of oxygen tension on kidney development in models designed to represent a pathological hypoxic insult. In vivo mouse models of moderate, transient, midgestational (12% O₂, 48 h, 12.5 dpc) or severe, acute, early-gestational (5.5-7.5% O₂, 8 h, 9.5-10.5 dpc) hypoxia were developed. The embryo itself is known to mature under hypoxic conditions with embryonic tissue levels of oxygen estimated to be 5%-8% (physiological hypoxia) when the mother is exposed to ambient normoxia. Both in vivo models generated phenotypes seen in patients with congenital anomalies of the kidney and urinary tract (CAKUT). Severe, acute, early hypoxia resulted in duplex kidney, while moderate, transient, midgestational hypoxia permanently reduced ureteric branching and nephron formation. Both models displayed hypoxia-induced reductions in β-catenin signaling within the ureteric tree and suppression of the downstream target gene, Ccnd1. Thus, we show a link between gestational hypoxia and CAKUT, the phenotype of which varies with timing, duration, and severity of the hypoxic insult.
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O