  • Search Results
  • Selective Halogen-Lithium Exchange of 1,2-Dihaloarenes for Successive [2+4] Cycloadditions of Arynes and Isobenzofurans.
Selective Halogen-Lithium Exchange of 1,2-Dihaloarenes for Successive [2+4] Cycloadditions of Arynes and Isobenzofurans.
Molecules (Basel, Switzerland) (2015-10-30)
Shohei Eda, Toshiyuki Hamura
ABSTRACT
Successive [2+4] cycloadditions of arynes and isobenzofurans by site-selective halogen-lithium exchange of 1,2-dihaloarenes were developed, allowing the rapid construction of polycyclic compounds which serve as a useful synthetic intermediates for the preparation of various polyacene derivatives.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%