  • Search Results
  • Shelf life, physicochemical, microbiological and antioxidant properties of purple cactus pear (Opuntia ficus indica) juice after thermoultrasound treatment.
Shelf life, physicochemical, microbiological and antioxidant properties of purple cactus pear (Opuntia ficus indica) juice after thermoultrasound treatment.
Ultrasonics sonochemistry (2015-07-19)
Nelly Del Socorro Cruz-Cansino, Esther Ramírez-Moreno, Jesús Ernesto León-Rivera, Luis Delgado-Olivares, Ernesto Alanís-García, José Alberto Ariza-Ortega, José de Jesús Manríquez-Torres, Diana Pamela Jaramillo-Bustos
ABSTRACT
The objective of this study was to evaluate changes in color, betalain content, browning index, viscosity, physical stability, microbiological growth, antioxidant content and antioxidant activity of purple cactus pear juice during storage after thermoultrasonication at 80% amplitude level for 15 and 25 min in comparison with pasteurized juice. Thermoultrasound treatment for 25 min increased color stability and viscosity compared to treatment for 15 min (6.83 and 6.72 MPa, respectively), but this last parameter was significantly lower (p<0.05) compared to the control and pasteurized juices (22.47 and 26.32 MPa, respectively). Experimental treatment reduced significantly (p<0.05) sediment solids in juices. Total plate counts decreased from the first day of storage exhibiting values of 1.38 and 1.43 logCFU/mL, for 15 and 25 min treatment, respectively. Compared to the control, both treatments reduced enterobacteria counts (1.54 logCFU/mL), and compared to pasteurized juice decreased pectinmethylesterase activity (3.76 and 3.82 UPE/mL), maintained high values of ascorbic acid (252.05 and 257.18 mg AA/L) and antioxidant activity (by ABTS: 124.8 and 115.6 mg VCEAC/100 mL; and DPPH: 3114.2 and 2757.1 μmol TE/L). During storage thermoultrasonicated juices had a minimum increase in pectinmethylesterase activity (from day 14), and exhibited similar total plate counts to pasteurized juice. An increase of phenolic content was observed after 14 days of storage, particularly for treatment at 80%, 25 min, and an increase in antioxidant activity (ABTS, DPPH) by the end of storage.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)