  • Search Results
  • Polarized Absorption Spectra of Single Crystals of Ion Radical Salts. I. Molecular Compounds of 7, 7, 8, 8-Tetracyano-p-quinodimethane with N, N, N', N'-Tetramethyl-p-phenylenediamine and N, N-Dimethyl-p-phenylenediamine.
Polarized Absorption Spectra of Single Crystals of Ion Radical Salts. I. Molecular Compounds of 7, 7, 8, 8-Tetracyano-<I>p</I>-quinodimethane with <I>N, N, N', N</I>'-Tetramethyl-<I>p</I>-phenylenediamine and <I>N, N</I>-Dimethyl-<I>p</I>-phenylenediamine.
Bulletin of the Chemical Society of Japan (1968)
Kuroda H, et al.