  • Search Results
  • Mechanism diversity in anodic oxidation of N, N-dimethyl-p-phenylenediamine by varying pH.
Mechanism diversity in anodic oxidation of <I>N, N</I>-dimethyl-<I>p</I>-phenylenediamine by varying pH.
Journal of Electroanalytical Chemistry (2013)
Maleki A and Nematollahi D.