  • Search Results
  • Spectrophotometric catalytic determination of an ultratrace amount of iron (III) in water based on the oxidation of N, N-dimethyl-p-phenylenediamine by hydrogen peroxide.
Spectrophotometric catalytic determination of an ultratrace amount of iron (III) in water based on the oxidation of <I>N, N</I>-dimethyl-<I>p</I>-phenylenediamine by hydrogen peroxide.
Analytical Chemistry (1988)
Hirayama K and Unohara N.