  • Search Results
  • N,N-dimethylformamide as a reaction medium for metal nanoparticle synthesis.
<I>N</I>,<I>N</I>-dimethylformamide as a reaction medium for metal nanoparticle synthesis.
Advances in Functional Materials (2009)
Pastoriza-Santos I and Liz-Marzan LM.