  • Search Results
  • Preparation of gold nanoparticles in formamide and N,N-dimethylformamide in the presence of poly (amidoamine) dendrimers with surface methyl ester groups.
Preparation of gold nanoparticles in formamide and <I>N</I>,<I>N</I>-dimethylformamide in the presence of poly (amidoamine) dendrimers with surface methyl ester groups.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects (2001)
Esumi K, et al.