  • Search Results
  • Measurements and molecular interactions for N,N-dimethylformamide with ionic liquid mixed solvents.
Measurements and molecular interactions for <I>N</I>,<I>N</I>-dimethylformamide with ionic liquid mixed solvents.
The Journal of Physical Chemistry B (2010)
Attri P, et al.