  • Search Results
  • The dynamics of liquid formamide, N-methylformamide, N,N-dimethylformamide, and N,N-dimethylacetamide. A dielectric relaxation study.
The dynamics of liquid formamide, <I>N</I>-methylformamide, <I>N</I>,<I>N</I>-dimethylformamide, and <I>N</I>,<I>N</I>-dimethylacetamide. A dielectric relaxation study.
Journal of Molecular Liquids (2002)
Barthel J, et al.