  • Search Results
  • Stability, permeability and growth-inhibitory properties of gonadotropin-releasing hormone liposaccharides.
Stability, permeability and growth-inhibitory properties of gonadotropin-releasing hormone liposaccharides.
Pharmaceutical research (2014-11-20)
Daryn Goodwin, Pegah Varamini, Pavla Simerska, Istvan Toth
ABSTRACT
In this study we aimed to address the poor drug-like properties of Gonadotropin-Releasing Hormone (GnRH) peptide through modification with lipids and carbohydrates. GnRH peptide was conjugated to 2-amino-D,L-octanoic acid (C8) and 2-amino-D,L-dodecanoic acid (C12) in monomer and dimer, along with (6-9) or without (2-5 and 11) a glucose moiety. Peptides were tested for their biological activity using different tumour cell lines. The toxicity of the constructs was evaluated in peripheral blood mononuclear cells (PBMC). All (glyco)lipopeptides showed improved metabolic stability in Caco-2 cell homogenates. Those with single lipid moiety (2, 4 and 8) exhibited prodrug-like properties. Permeability across Caco-2 cell monolayers was enhanced in the dimer C8-modified (glyco)lipopeptide (3) and the lipopeptide with C12 inserted mid-sequence (11). Most of the constructs showed moderate-to-high antiproliferative activity against GnRH-receptor positive DU145 and OVCAR-3 cells (up to 60%). Compound 11 was the most effective with IC50 = 26.4 ± 1.07 μg.ml(-1), which was comparable to triptorelin (25.1 ± 1.14 μg.mL(-1)). The sensitivity of OVCAR-3 cells to the effect of all analogues except for 11 decreased significantly in estrogen-reconstituted media. Only compounds 2, 4, 5 and 8 showed a steroid-dependent effect in DU145 cells. No compounds exhibited significant toxicity on PBMCs. These results indicated lipidation and glycosylation improves the druggability of GnRH and could lead to an increased direct antitumour activity in some hormone dependent and independent reproductive cancers.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
HEPES
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)