  • Search Results
  • Enhancement of the oral bioavailability of breviscapine by nanoemulsions drug delivery system.
Enhancement of the oral bioavailability of breviscapine by nanoemulsions drug delivery system.
Drug development and industrial pharmacy (2014-08-13)
Yan Ma, HaiGang Li, ShiXia Guan
ABSTRACT
Aim to design an effective breviscapine nanoscale drug delivery system to realize the improvement of its oral bioavailability. Based on the investigations of the stabilities in the gastrointestinal tract (GIT), permeation and efflux across the cell membrane, the breviscapine nanoemulsion (NE) was formulated and evaluated in vitro and in vivo. The globule size and polydispersity index of the NE was 45.6 nm and 0.105, and the efficient encapsulation was 95.2%. In vitro, the drug release from NEs in pH 6.8 PBS fit to the first-order kinetics. The Caco-2 cell transport experiments showed that the breviscapine NE facilitated the improvement of the apparent permeability coefficient (Papp) from the apical side to basilar side compared with the free drug. In vivo, the relative bioavailability of breviscapine NE reached to 249.7%. All the studies implicated that the NE carrier contributed to the enhancement of the oral absorption of breviscapine due to the improved stability and permeation in the GIT. The nanoemulsions technology is better for the poor permeable and unstable active agents in GIT as well as helps the industrial scale process.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, analytical standard