MilliporeSigma
Glycosidase inhibitors from the roots of Glyphaea brevis.
Phytochemistry (2014-12-04)
Diane Patricia Apie Gossan, Abdulmagid Alabdul Magid, Philomène Akoua Kouassi-Yao, Jean-Bernard Behr, Antoine Coffy Ahibo, Léon Atoutou Djakouré, Dominique Harakat, Laurence Voutquenne-Nazabadioko
ABSTRACT
Ten phenylalkyl-substituted iminosugars (1-10) and a cinnamic acid derived glucoside (11) were isolated from the roots of Glyphaea brevis (Malvaceae). Their structures were elucidated by 1D and 2D NMR analysis, as well as by HR-ESIMS. Compounds 1-10 retain an unprecedented structure composed of an iminosugar-like core identified as 1-deoxyfuconojirimycin in glyphaeaside A1-A4 (1, 2, 5, 6), 1-deoxygalactonojirimycin in glyphaeaside B1-B5 (3, 4, 7-9) or 1-deoxynojirimycin in glyphaeaside C (10), substituted by a β-d-glucopyranose in compounds 2, 4, 6 and 9. These compounds feature a di-, tri- or tetra-hydroxylated nine-carbon chain at the pseudo-anomeric position, substituted by a terminal phenyl group. All alkyl C-iminosugars displayed potent and selective inhibition towards β-glucosidase with IC50 values ranging from 0.15 to 68μM. Compound 10 with an 1-deoxynojirimycin backbone was the most active and was found to act as a competitive inhibitor with Ki=31nM, therefore emerging as one of the most potent inhibitor of β-glucosidase reported to date. Inhibition of β-mannosidase was observed with compounds 1, 3, 7 and 10, but only weak inhibition could be detected with the alkyl-C-iminosugars on the other tested glycosidases (α-glucosidase, α-fucosidase, α- and β-galactosidase).
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC