  • Search Results
  • Hyaluronan deposition and co-localization with inflammatory cells and collagen in a murine model of fungal allergic asthma.
Hyaluronan deposition and co-localization with inflammatory cells and collagen in a murine model of fungal allergic asthma.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-02-13)
Sumit Ghosh, Amali E Samarasinghe, Scott A Hoselton, Glenn P Dorsam, Jane M Schuh
ABSTRACT
Allergic asthma is a chronic inflammatory disease of the airways characterized by excessive inflammation and remodeling of the extracellular matrix (ECM) and associated cells of the airway wall. Under inflammatory conditions, hyaluronan (HA), a major component of the ECM, undergoes dynamic changes, which may in turn affect the recruitment and activation of inflammatory cells leading to acute and chronic immunopathology of allergic asthma. In the present study, we measured the changes in HA levels generated at sites of inflammation, and examined its effect on inflammatory responses and collagen deposition in an Aspergillus fumigatus murine inhalational model of allergic asthma. We found that HA levels are elevated in allergic animals and that the increase correlated with the influx of inflammatory cells 5 days after the second allergen challenge. This increase in HA levels appeared largely due to upregulation of hyaluronidase-1 (HYAL1) and hyaluronidase-2 (HYAL2). Furthermore, HA co-localizes with areas of new collagen synthesis and deposition. Overall, our findings contribute to the growing literature that focuses on the components of ECM as inflammatory mediators rather than mere structural support products. The evidence of HA localization in fungal allergic asthma provides the impetus to study HA more closely with allergic leukocytes in murine models. Further studies examining HA's role in mediating cellular responses may help to develop targets for treatment in patients with severe asthma due to fungal sensitization.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology