  • Search Results
  • Accelerated onset of senescence of endothelial progenitor cells in patients with type 2 diabetes mellitus: role of dimethylarginine dimethylaminohydrolase 2 and asymmetric dimethylarginine.
Accelerated onset of senescence of endothelial progenitor cells in patients with type 2 diabetes mellitus: role of dimethylarginine dimethylaminohydrolase 2 and asymmetric dimethylarginine.
Biochemical and biophysical research communications (2015-02-24)
Qiong Yuan, Chang-Ping Hu, Zhi-Cheng Gong, Yong-Ping Bai, Si-Yu Liu, Yuan-Jian Li, Jun-Lin Jiang
ABSTRACT
The risk of cardiovascular complications in diabetic patients is mainly associated with endothelial dysfunction. Reduced number of EPCs and impaired function of EPCs in diabetes result in imbalance of endothelial homeostasis and dysfunction of vessels. In patients with diabetes mellitus, plasma levels of asymmetric dimethylarginine (ADMA) were elevated, while the expression and activity of dimethylarginine dimethylaminohydrolase (DDAH) were reduced. In the present study, we investigated the role of the DDAH2/ADMA pathway in the senescence of EPCs in type 2 diabetic patients and cultured EPCs treated with high glucose. The results showed that the percentage of senescent EPCs increased while the expression of DDAH2 decreased concomitantly with an increase in the plasma levels of ADMA in patients with type 2 diabetes mellitus (T2DM). Similar results were seen in cultured EPCs treated with high glucose. Exogenous application of ADMA accelerated the senescence of EPCs in a dose-dependent manner, and overexpression of DDAH2 inhibited high glucose-induced EPCs senescence. In addition, it has also been reported that DDAH/ADMA pathway is regulated by silent information regulator 1 (SIRT1) in endothelial cell. In the present study, we found decreased expression of SIRT1 both in T2DM patients and EPCs pretreated with high glucose. And resveratrol (activating SIRT1) inhibited high glucose-induced EPCs senescence by upregulating the expression of DDAH2 and decreasing the levels of ADMA. Taken together, we concluded that DDAH2/ADMA is involved in the accelerated senescence of EPCs in diabetes, which is associated with the activation of SIRT1.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard