  • Search Results
  • Nanoparticle biphasic calcium phosphate loading on gelatin-pectin scaffold for improved bone regeneration.
Nanoparticle biphasic calcium phosphate loading on gelatin-pectin scaffold for improved bone regeneration.
Tissue engineering. Part A (2015-01-21)
Thuy Ba Linh Nguyen, Young Ki Min, Byong-Taek Lee
ABSTRACT
Biphasic calcium phosphate (BCP) nanoparticles were loaded with porous gelatin-pectin (GE-P) scaffolds. The biodegradable gelatin-pectin-BCP scaffolds were produced as miscible mixtures with well-defined interconnected pores to facilitate osteoconductivity and enhance bone formation. It was observed that the compressive strength increased with the loading of BCP nanoparticles. From in vitro results, cell adhesion, viability, and proliferation were found in the GE-P-10 scaffolds in comparison with those without BCP, resulting in high alkaline phosphate (ALP), and osteopontin (OPN) expression at 21 days. Micro-computed tomography data, hematoxylin and eosin staining, and immunohistochemistry (OPN, OCN, COL I, and COL II) confirmed rapid new bone formation in rabbit models. Our results provide a novel and simple method to provide an adequate scaffold, and thus GE-P-BCP porous scaffolds may be appropriate candidates for bone tissue engineering.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
3-Benzidino-6-(4-chlorophenyl)pyridazine, ≥98% (HPLC), solid
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)