  • Search Results
  • An inverse association between magnesium in 24-h urine and cardiovascular risk factors in middle-aged subjects in 50 CARDIAC Study populations.
An inverse association between magnesium in 24-h urine and cardiovascular risk factors in middle-aged subjects in 50 CARDIAC Study populations.
Hypertension research : official journal of the Japanese Society of Hypertension (2014-10-31)
Yukio Yamori, Miki Sagara, Shunsaku Mizushima, Longjian Liu, Katsumi Ikeda, Yasuo Nara
ABSTRACT
Serum, plasma and dietary magnesium (Mg) have been reported to be inversely associated with cardiovascular disease risk factors. We examined the associations between the 24-h urinary Mg/creatinine (Cre) ratio and cardiovascular disease risk factors, such as body mass index (BMI), blood pressure (BP), serum total cholesterol (TC) and prevalence of obesity, hypertension and hypercholesterolemia. A cross-sectional analysis was conducted among 4211 participants (49.7% women) aged 48-56 years in 50 population samples from 22 countries in the World Health Organization-coordinated Cardiovascular Diseases and Alimentary Comparison (CARDIAC) Study (1985-1994). In linear regression analyses, Mg/Cre ratio was inversely associated with BMI, systolic BP (SBP), diastolic BP (DBP) and TC (P for linear trend <0.001 for each). These associations were not markedly altered by adjustment for traditional risk factors, urinary markers or cohort effects. Multivariate-adjusted mean values for the subjects in the highest Mg/Cre ratio quintile were 6.3, 3.4, 5.3 and 4.6% lower than those for the subjects in the lowest quintile for BMI, SBP, DBP and TC (P < 0.001, respectively). The prevalence of obesity, hypertension and hypercholesterolemia was 2.10 (95% confidence interval: 1.50, 2.95), 1.55 (1.25, 1.92) and 2.06 (1.63, 2.62) times higher (P < 0.001, respectively) among the subjects in the lowest Mg/Cre ratio quintile than in the subjects in the highest quintile. These associations were not appreciably altered by adjustment for potential confounding variables. In conclusion, higher 24-h urinary Mg/Cre ratio was associated with lower cardiovascular disease risk factors, including BMI, BP, TC, obesity, hypertension and hypercholesterolemia.
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Dibutyl phthalate, Selectophore
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)