  • Search Results
  • Carthamus, Salvia and Stachys species protect neuronal cells against oxidative stress-induced apoptosis.
Carthamus, Salvia and Stachys species protect neuronal cells against oxidative stress-induced apoptosis.
Pharmaceutical biology (2014-09-23)
Marjan Tavakkoli, Ramin Miri, Amir Reza Jassbi, Nasrollah Erfani, Mojtaba Asadollahi, Mansoureh Ghasemi, Luciano Saso, Omidreza Firuzi
ABSTRACT
Finding effective therapies for neurodegenerative diseases is of utmost importance for the aging population. Plants growing in Iran are rich sources of antioxidants and active phytochemicals. The protective capacity of plants, with a special focus on those with reported antioxidant or neuroprotective potential or nervous system-related applications in folk medicine, was tested against oxidative stress-induced apoptosis. Aerial parts of 20 plants including Carthamus, Salvia, and Stachys species were extracted with 80% methanol and dichloromethane and preincubated with neuronal PC12 cells for 3 h. Oxidative stress and apoptosis were induced by hydrogen peroxide (75 µM, 1 h exposure). Cell viability and intracellular reactive oxygen species (ROS) were measured by MTT and 2',7'-dichlorofluorescein-diacetate (DCFH-DA) assays, respectively, while apoptosis was determined by annexin V-FITC/propidium iodide staining by a flow cytometer. Eighty percent methanol extracts of Carthamus oxyacantha Bieb. (Asteraceae), Salvia santolinifolia Boiss. (Lamiaceae), and Salvia sclarea L. (Lamiaceae) at the concentration of 100 μg/ml showed significant neuroprotection in the MTT assay by 38.7, 34.7, and 39.5%, respectively, and inhibited intracellular ROS by 48.6, 61.9, and 61.4%, respectively. The first two extracts also significantly inhibited apoptosis. Dichloromethane extracts of C. oxyacantha and Stachys pilifera Benth. (Lamiaceae) at the concentration of 25 μg/ml showed neuroprotection by 27.5 and 26.5%, respectively, and inhibited ROS by 44.5 and 39.4%, respectively. The above-mentioned plants seem to have important biological activities and their further study may lead to the discovery of new natural therapeutics useful against disorders such as Alzheimer and Parkinson diseases.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Methanol, analytical standard
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)