  • Search Results
  • Intracellular delivery of cytochrome c by galactosylated albumin to hepatocarcinoma cells.
Intracellular delivery of cytochrome c by galactosylated albumin to hepatocarcinoma cells.
Journal of drug targeting (2014-04-16)
Tzyy-Harn Yeh, Fe-Lin Lin Wu, Li-Jiuan Shen
ABSTRACT
In some cancer cells, translocation of cytochrome c (Cyt c) from mitochondria to the cytoplasma is inhibited. This inhibition prevents cells from undergoing apoptotic cell death and can lead to uncontrolled cell growth. Increasing cytoplasmic concentration of Cyt c can induce apoptosis in cancer cells as a strategy of cancer therapy. Here we proposed a galactosylated albumin based carrier for intracellular delivery of Cyt c to hepatocarcinoma cells. Galactosylated albumin is recognized by highly expressed asialoglycoprotein receptors (ASGPR) on hepatocarcinoma cells and is further internalized into cells via receptor mediated endocytosis. Cyt c was chemically conjugated to galactosylated albumin with a reducible disulfide linker in order to release Cyt c from the carrier inside cells. We tested cellular uptake and cytotoxicity of Cyt c conjugates in ASGPR positive and negative hepatocarcinoma cells. The results showed galatosylated albumin significantly increased cellular uptake in both cell types resulting in cytotoxicity in a dose dependent manner through the induction of apoptosis. The lack of ASGPR specific uptake might be due to other carbohydrate-recognizing receptors expressed on tumor cells. In general, our work has shown that intracellular delivery of Cyt c to tumor cells can be an alternative therapeutic approach and galactosylated albumin can be a protein drug carrier for intracellular delivery.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)