  • Search Results
  • Study of chemical pretreatment and enzymatic saccharification for producing fermentable sugars from rice straw.
Study of chemical pretreatment and enzymatic saccharification for producing fermentable sugars from rice straw.
Bioprocess and biosystems engineering (2013-12-19)
Wen-Hsing Chen, Yi-Chun Chen, Jih-Gaw Lin
ABSTRACT
This study evaluated a cost-effective approach for the conversion of rice straw into fermentable sugars. The composition of rice straw pretreated with 1 % sulfuric acid or 1 % sodium hydroxide solution was compared to rice straw with no chemical pretreatment. Enzymatic saccharification experiments on non-pretreated rice straw (NPRS), pretreated rice straw (PRS), and pretreated rice straw with acid hydrolysate (PRSAH) were conducted in a series of batch reactors. The results indicated that pretreating the rice straw with dilute acid and base increased the cellulose content from 38 % to over 50 %. During enzymatic saccharification, straight aliphatic cellulose was hydrolyzed before branched hemicellulose, and glucose was the major hydrolysis product. The glucose yield was 0.52 g glucose/g for NPRS and was comparable to the yields of 0.50 g glucose/g for PRS and 0.58 g glucose/g for PRSAH. The hydrolysis of rice straw to produce glucose can be described by a first-order reaction with a rate constant of 0.0550 d(-1) for NPRS, 0.0653 d(-1) for PRSAH, and 0.0654 d(-1) for PRS. Overall, the production of fermentable sugars from ground rice straw will be more cost effective if the straw is not pretreated with chemicals.
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard